Cole-Parmer Essentials Write-On Tape, White, 3/4" x 14 yds, 4/pk
Representative image only.
Cole-Parmer Essentials Item # RK-06530-31
Identify specific applications by color coding
  • Withstands temperatures from –10 to 250 ºF (–23 to 121 ºC)
  • Waterproof, and oil and acid resistant
PRODUCT OPTIONS
$64.09CAD / Pkg of 4
BUY MORE & SAVE
  • 1-5$64.09CAD / Pkg of 4
  • 6-29$57.68CAD / Pkg of 4
  • 30-119$54.48CAD / Pkg of 4
  • 120+$51.27CAD / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • ColorWhite
  • Width (in)3/4
  • Length (ft)42
  • Width (cm)1.9
  • Length (m)12.8
  • DescriptionWrite-On Tape, White, 3/4\" x 14 yds, 4/pk
Key Features
  • Withstands temperatures from –10 to 250 ºF (–23 to 121 ºC)
  • Waterproof, and oil and acid resistant
More About this Item
Use tape to identify specific applications by color coding. This durable labeling tape is waterproof as well as oil and acid resistant. Tape can be used with pencil, pen, or permanent marker.
Use tape to identify specific applications by color coding. Durable labeling tape is waterproof, and oil and acid resistant. Tape can be used with pencil, pen, or permanent marker. Withstands temperatures from –10 to 250ºF (–23 to 121ºC).