Cole-Parmer Essentials Water Bath Clamp, Three-Prong, Large, 1.81" Max Grip, 0.43" Max Tank Wall
Representative image only.
Cole-Parmer Essentials Item # RK-08043-10
Provide support for your specific laboratory equipment
  • 1 year warranty
PRODUCT OPTIONS
$60.41CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeEquipment, Water Bath
  • Min Grip Range (in)0
  • Max Grip Range (in)1.81
  • Length (in)4.61
  • DescriptionWater Bath Clamp, Three-Prong, Large, 1.81\" Max Grip, 0.43\" Max Tank Wall
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These C-shaped clamps with thumbscrew attach securely to your bath wall.
Three-prong clamp has two adjustment screws for greater flexibility over a wider range. Three prong clamp includes both slip-on vinyl and fiberglass sleeves.