Cole-Parmer Viscosity Cup #3, 150 to 790 cSt, with NIST-Traceable Report
Representative image only.
Cole-Parmer Item # RK-08700-67
Simple and convenient – no complicated training needed
PRODUCT OPTIONS
$486.59CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Viscosity150 to 790 centistokes
  • Cup Number3
  • Nist-TraceableYes
  • DescriptionViscosity Cup #3, 150 to 790 cSt, w/NIST-traceable report
More About this Item
Measure the viscosity of liquids such as paint, lacquer, oil, and syrups. Simply insert the cup into your sample and lift the viscosity cup out, starting a stopwatch/timer. Cup is made of series 400 stainless steel and requires 44 mL of sample for the test. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. A conversion sheet is supplied that will convert the Zahn time to a centistokes value.
Cup includes an NIST-traceable calibration report supplied by the manufacturer to meet your quality program requirements.