DescriptionMid-Size UV Viewing Cabinet for 8-watt UV Lamps, 115VAC
Warranty1 Year
Includes
Built-in overhead white light.
Key Features
Stay comfortable with soft rubber viewport viewing area
Removable bottom panel allows cabinet to be placed over large samples or transilluminator
Block out ambient light with lightweight access curtain
Protect your eyes with contrast control filter
Two lamps illuminate cabinet's interior from two sides
Lamps are removable for use in other applications
1 year warranty
More About this Item
These customizable UV viewing cabinets provide a compact darkroom environment for viewing materials under ultraviolet conditions. Manufactured of durable lightweight plastic, the cabinets are designed for long life and rugged use in a wide range of applications. Bottom panel of each cabinet is removable and the unit can easily be placed over samples or a transilluminator. A lightweight access curtain allows you easy entry and blocks out external light. The view port combines a comfortable soft rubber, which can accommodate eyeglasses, and a contrast control filter blocks out ultraviolet radiation. A wide range of cabinet models and ultraviolet wavelength configurations for use in the laboratory or industrial applications are available.
Mid-size UV viewing cabinet is designed to accommodate one or two 8-watt ultraviolet lamps (sold separately). Lamps plug into the service outlet located on top of the cabinet and can be placed over the cabinet opening with the filter glass facing into the cabinet. Using two lamps provides uniform illumination from two sides of the cabinet. Lamps are removable for use in other applications. A rocker switch is located on the back of the lamp. The cabinet master ON/OFF rocker switch must be on to operate the white light and service outlets. Cabinet also feature an interior overhead white light for basic sample illumination.
Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.
Warning: Do not expose eyes and skin to shortwave ultraviolet radiation as rays may be harmful to unprotected eyes and skin.