This item is now discontinued. Please browse similar items
Cole-Parmer Essentials Versatile Carboy Cap, 120 mm, Open with Solid Adapter; 2/Pk
Representative image only.
Cole-Parmer Essentials Item # RK-06059-29
Leakproof caps with the flexibility of interchangeable ports
  • Finger grips allow easy tightening and opening
  • Choose from a variety of sizes and configurations*
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPP
  • Cap / Stopper Size120 mm
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing IDOpen with Solid Adapter
  • DescriptionVersatile Carboy Cap, 120 mm, Open with Solid Adapter; 2/Pk
Key Features
  • Finger grips allow easy tightening and opening
  • Choose from a variety of sizes and configurations*
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
More About this Item
Versatile carboy caps are made of autoclavable polypropylene and come in industry-standard sizes.
Solid caps do not accept insert adapters.