Cole-Parmer VapLock™ Port Plug, Natural PTFE, 1/4"-28 UNF(M); 10/PK
Representative image only.
Cole-Parmer Item # RK-12020-47
Use plugs to seal unused ports for a leak-resistant system
  • Seal unused manifold and bottle cap ports
PRODUCT OPTIONS
$48.03CAD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPTFE
  • Connection TypeUNF(M)
  • Thread size UNF(M) (port 1) (in)1/4\"-28
  • ColorWhite
  • DescriptionPort Plug, 1/4\"-28 UNF(M), natural PTFE; 10/pk
Key Features
  • Seal unused manifold and bottle cap ports
More About this Item
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.