Cole-Parmer VapLock™ Plug, Stainless Steel, Luer Plug x 1/32" UNF(M); 1/Ea
Representative image only.
Cole-Parmer Item # RK-12031-87
Use plugs to seal unused ports for a leak-resistant system
  • Seal unused manifold and bottle cap ports
PRODUCT OPTIONS
$76.91CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeUNF(M)
  • Thread size UNF(M) (port 1) (in)1/3\"
  • ColorGrey
  • DescriptionPlug, Stainless Steel, Luer Plug x 1/32\" UNF(M); 1/Ea
Key Features
  • Seal unused manifold and bottle cap ports
More About this Item
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.