This item is now discontinued. Please browse similar items
Spex VapLock™ Fitting, Polypropylene, Elbow, Hose Barb Union, 5/32" ID; 10/PK
Representative image only.
Spex Item # RK-12023-67
Connect soft tubing using barbed fittings
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Number Of Ports2
  • Connection TypeBarb
  • Barb Size / Tubing Id (Port 1) (mm)3.97
  • Barb Size / Tubing Id (Port 1) (in)5/32
  • Barb Size / Tubing Id (Port 2) (mm)3.97
  • Barb Size / Tubing Id (Port 2) (in)5/32
  • ColorClear
  • Description Fitting, Polypropylene, Elbow, Hose Barb Union, 5/32\" ID; 10/PK
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
More About this Item
These elbow barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.