Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 20 L; 1/ea

Representative image only.
Mfr # 392-2004-OEM – Item # RK-06059-99
Safely transport hazardous liquids
  • UN/DOT rated for Group II and hazardous chemicals
  • Large top handle provides a comfortable full-hand grip
  • HDPE construction offers excellent chemical compatibility

PRODUCT OPTIONS

$55.99CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (L)20
  • MaterialHDPE
  • Cap MaterialHDPE
  • Cap Size (mm)70
  • Overall Length (in)11 1/4
  • Overall Width (in)9
  • Overall Height (in)15 1/2
  • DescriptionWaste Container, UN/DOT, S70 cap; 20 L; 1/ea
ISO9001 Registered SupplierMade in the U.S.A.

Key Features

  • UN/DOT rated for Group II and hazardous chemicals
  • Large top handle provides a comfortable full-hand grip
  • HDPE construction offers excellent chemical compatibility

More About this Item

Sturdy, rectangular UN/DOT carboys are easy to fill and ship for disposal. Safely use these single-use carboys to transfer and store liquid waste, use as waste traps with automated instruments, or as part of your HPLC system.

UN rating: 3H1/Y1.8/100

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat