This item is now discontinued. Please browse similar items
Cole-Parmer Twist-Lock Adapter without Guard, 3/4"NPT(M); PVDF
Representative image only.
Cole-Parmer Item # RK-05993-86
For a complete system, order an electrode, cable, and in-line adapter – it’s as easy as 1-2-3!
  • A stop pin is installed in the adapter to assure positive electrode positioning
  • A 1/4 turn is all that is required for proper sensor installation into the adapter
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPVDF
  • Compatible WithCole-Parmer Twist-Lock electrode and 3/4\" NPT(F) pipe tee
  • Connections3/4\" NPT(M) to Twist-Lock adapter
  • DescriptionTwist-Lock Adapter without Guard, 3/4\"NPT(M); PVDF
  • Warranty1 year
1 Year Warranty
Key Features
  • A stop pin is installed in the adapter to assure positive electrode positioning
  • A 1/4 turn is all that is required for proper sensor installation into the adapter
  • 1 year warranty
More About this Item
Pipe tee adapters are specially designed to accept Twist-Lock probes, come with 3/4" process connection threads, without an integrated probe guard, and are composed of PVDF.
Other adapters are available with guards and composed of stainless steel for higher temperature use. Choose the in-line adapter that suits your needs—stainless steel adapters are suggested for temperatures above 176°F (80°C).