Cole-Parmer Tuff-Tip pH Electrode, No ATC, BNC, 3/4", 10 ft Cable
Representative image only.
Cole-Parmer Item # RK-27020-00
Less chance of breakage
  • Don't worry about bulb breakage
  • Eliminate electrical noise with graphite-tipped models
  • 1 year warranty
PRODUCT OPTIONS
$391.92CAD / Each
BUY MORE & SAVE
  • 1-5$391.92CAD / Each
  • 6-11$352.73CAD / Each
  • 12-23$333.13CAD / Each
  • 24+$313.54CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode Body MaterialKynar®
  • Reference JunctionCeramic
  • Max Temperature (° C)110
  • Temperature CompensationNo
  • Thread Size NPT(M) (in)3/4
  • ATC ConnectorNone
  • ATC ElementNone
  • ConnectorBNC
  • DescriptionTuff-Tip pH Electrode, No ATC, BNC, 3/4\", 10 ft Cable
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Don't worry about bulb breakage
  • Eliminate electrical noise with graphite-tipped models
  • 1 year warranty
More About this Item
The unique Tuff-Tip design helps protect bulbs. Choose chemical-resistant Ryton® PPS standard electrode or with graphite tip. Ideal for viscous or severe applications, these electrodes reduce contamination and clogging with uniform pore size.