Cole-Parmer Essentials Tight-Ties Nylon Cable/Zip Ties, 8"; 100/PK
Representative image only.
Cole-Parmer Essentials Item # RK-98766-46
Chemical-resistant and corrosion-proof fasteners
  • Nylon is corrosion-proof, chemical resistant, and non-conductive
  • Tensile strength is 50 lbs
$16.94CAD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)8
  • ColorWhite
  • MaterialNylon
  • DescriptionTight-Ties Nylon Cable/Zip Ties, 8\"; 100/Pk
1 Year Warranty
Key Features
  • Nylon is corrosion-proof, chemical resistant, and non-conductive
  • Tensile strength is 50 lbs
More About this Item
Use to fasten pressure, vacuum, and liquid tubing lines or bundle lines together. Use from -40 to 185°F (-40 to 85°C). When fastened, diameters adjust from 5/32" to 4" (2.6 to 10.2 cm).