This item is now discontinued. Please browse similar items
Cole-Parmer ThinTouch™ Nitrile Gloves, 12 Boxes/Kit (6 Medium, 6 large)
Representative image only.
Cole-Parmer Item # RK-25000-19
Mixture of different sizes for your convenience
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Superb dexterity and elasticity
  • Soft feel provides comfort for extended wear
  • Each case contains 12 boxes of gloves, each box containing 100 gloves
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5.1
  • SizeMedium/Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffBeaded
  • SterileNo
  • Includes6 boxes of Medium, 6 boxes of Large
  • PowderedNo
  • DescriptionThinTouch™ Nitrile Gloves, Medium to Large; 12 Boxes/Kit
Includes
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Superb dexterity and elasticity
  • Soft feel provides comfort for extended wear
  • Each case contains 12 boxes of gloves, each box containing 100 gloves
More About this Item
These powder-free, nitrile gloves offer a soft feel and textured fingertips for a better grip. Their beaded cuffs make them easy to put on and take off. Disposable gloves, for single-use only. Ambidextrous.