This item is now discontinued. Please browse similar items
Foxx Life Sciences Syringe Filter, 13 mm, 0.45 um PTFE; 100/Pk
Representative image only.
Cole-Parmer Mfr # 386-3116-OEM – Item # RK-06060-77
Economical, single use, and efficient filtering stream for high output filtration from viscous and particle-laden solutions
  • Reduce bursting with reinforced polypropylene housing Take advantage of a large effective filtration area Standard female luer-lock inlet and male luer-slip outlet Available in various membrane size, pore size, and material combinations
SPECS & DESCRIPTION
Specifications & Description
  • Membrane Materialhydrophilic PTFE
  • Pore Size (µm)0.45
  • Membrane Diameter (mm)13
  • Housing MaterialPP
  • Connectionsfemale luer-lock inlet and male luer-slip outlet
  • DescriptionSyringe filter, 13 mm, .45 um hydrophilic PTFE; 100/pk
ISO9001 Registered Supplier
Key Features
  • Reduce bursting with reinforced polypropylene housing Take advantage of a large effective filtration area Standard female luer-lock inlet and male luer-slip outlet Available in various membrane size, pore size, and material combinations
More About this Item
Use the single-use syringe filters for clarification and prefiltration, and even for removing fine particulate with the membrane’s large effective filtration area. Reinforced polypropylene housing has secure connections for male luer slip outlet and female luer lock inlet. Non-sterile bulk pack of syringe filters 100 per pack. The membrane type and pore size are visible on each filter housing as well.