This item is now discontinued. Please browse similar items
Cole-Parmer Submarine Gel System Additional Tray for 28561-00
Representative image only.
Cole-Parmer Item # RK-28561-50
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSubmarine Gel System Additional Tray for 28561-00
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Gel Tray lets you pour gels in the unit using the tapered baffles, outside of the unit by taping the ends of the tray; or outside of the unit using the gel casters sold seperately. Tray is UV-transparent acrylic. Multiple comb slots let you cast gels for simultaneous short runs. With 6 comb slots; pathlength of 2.6 cm/slot