This item is now discontinued. Please browse similar items
Cole-Parmer Sterile PP Culture Tubes w/ 2-Position PE Caps; 17x100mm, Natural; 500/Bx
Representative image only.
Cole-Parmer Item # RK-63600-20
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPE
  • Length (mm)100
  • Depth (mm)100
  • Diameter (mm)17
  • DescriptionSterile PP Culture Tubes w/ 2-Position PE Caps; 17x100mm, Natural; 500/Bx
Sterile
More About this Item
Heavy-walled tubes resist breakage. Choose from clear polystyrene (PS) or natural polypropylene (PP). The 12 x 75 mm tubes have a volume of 6 mL; the 17 x 100 mm tubes have a volume of 16 mL. Tubes are sterile; include two-position PE caps.