This item is now discontinued. Please browse similar items
Cole-Parmer StableTemp Double Block for Titer Plates
Representative image only.
Cole-Parmer StableTemp Item # RK-36600-27
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Block Number Of Openings20
  • Height (in)2 1/4
  • Width (in)3 3/4
  • Length (in)6
  • DescriptionDouble Block for Titer Plates
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Titer Plate Block is a double size block. Block holds a standard Microplate and is recessed for better stability Flat surface can hold flat or round bottom plates. Double block fits 2, 4, or 6 block heaters.
Block holds a standard microplate. Flat surface can hold flat or round bottom plates. Double plate fits 2, 4, or 6 block heaters