Cole-Parmer Spex® 2662 Homogenizer Cryo-Block, Aluminum, 24x5mL PC Vials 04576-98; 2/PK
Representative image only.
Cole-Parmer Item # RK-04575-85
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Enables high-throughput cryogenic grinding
  • Increases yields by grinding samples at sub-zero temperatures
  • Each pair holds up to 48 tubes for rapid and efficient sample preparation
PRODUCT OPTIONS
$747.75CAD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)5
  • Tubes Held24
  • MaterialAluminum
  • DescriptionHomogenizer Cryo-Block, Aluminum, 24 x 5 mL PC Vials 04576-98; 2/PK
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Enables high-throughput cryogenic grinding
  • Increases yields by grinding samples at sub-zero temperatures
  • Each pair holds up to 48 tubes for rapid and efficient sample preparation
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.
Each aluminum cryo-block holds 24 (5 mL) polycarbonate vials 04576-98. Chill cryo-blocks in the cryo-station 04575-81 to maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis. The blocks preserve pesticides, RNA, and proteins for heat sensitive extraction methods. Use two at a time in the clamp assembly 04605-08 to keep it in balance.