This item is now discontinued. Please browse similar items
Cole-Parmer SODIUM Low Level Sodium Electrode, S7 connector
Representative image only.
Cole-Parmer Item # RK-94785-04
Accurately monitor sodium levels in the ppb range
  • Ideal for power plant monitoring of steam turbines and stainless steel generators
  • Sodium ion range: 0.1 ppb to 10 ppm
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Housing MaterialGlass, combination
  • DescriptionLow Level Sodium Electrode, S7 connector
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for power plant monitoring of steam turbines and stainless steel generators
  • Sodium ion range: 0.1 ppb to 10 ppm
  • 1 year warranty
More About this Item
Economical alternative to accurately detect sodium ion impurities in the ppb range. Early detection helps to prevent sodium ion stress, corrosion, and cracking. Glass body combination electrode has an ammonia chloride reference solution. Rlectrode is avaliable with a BNC connector or a S7/detachable lead connection.
Model 94785-04 comes with an S7 connector. Order 3-ft (1-m) cable with CD/SN6-to-male BNC connector 05659-12 separately.