This item is now discontinued. Please browse similar items
Cole-Parmer Essentials Sieve; No 25, Half-Height, 12", SS/SS, NIST-Traceable Calibration Certificate
Representative image only.
Cole-Parmer Item # RK-59995-57
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)25
  • Tyler Standard Screen Size (mesh)24
  • Sieve Opening (mm)0.707
  • Outside Diameter (in)12
  • Height (InchShort)1.625
  • DescriptionSieve; No 25, Half-Height, 12\", SS/SS, NIST Cal Cert
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.