Cole-Parmer Self-Supportng Single Arm Gooseneck Fiber Optic Light Pipe, 20" (50.8 cm)
Representative image only.
Cole-Parmer Item # RK-41720-75
No support needed
  • Flexible arms offer positioning versatility
  • Idea for use with microscopes and sided illumination of a single sample
  • 1 year warranty
PRODUCT OPTIONS
$546.63CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Light Pipe Length (in)20
  • Light Pipe Outside Dimension (OD) (in)0.56
  • Light Pipe Length (cm)50.8
  • Light Pipe Outside Dimension (OD) (cm)1.4224
  • DescriptionSelf-Supporting Light Pipe Fiber Optic Single
  • Warranty1 Year
1 Year Warranty
Key Features
  • Flexible arms offer positioning versatility
  • Idea for use with microscopes and sided illumination of a single sample
  • 1 year warranty
More About this Item
Self-supporting light pipes require no stand or additional support. They connect directly to the illuminator. Flexible gooseneck is constructed of glass fibers sheathed in a steel monocoil.
Illumination base (41720-40) sold separately.