This item is now discontinued. Please browse similar items
Cole-Parmer Scientific Technical Pens, 0.2 mm Red; 6/Pk
Representative image only.
Cole-Parmer Item # RK-98766-17
Choose the color and tip size to meet your lab writing needs
  • Record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams
  • Select from black, blue, green, red, or an assortment of these colors
SPECS & DESCRIPTION
Specifications & Description
  • Tip Style0.2 mm
  • Pen ColorRed
  • DescriptionScientific Technical Pens, 0.2 mm Red; 6/Pk
Key Features
  • Record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams
  • Select from black, blue, green, red, or an assortment of these colors
More About this Item
Write on any smooth surface; ideal for plasticware, glassware, photos, slides, and more. Waterproof, nonfading ink dries instantly to prevent smearing and bleeding. Extra-fine, plastic writing tip stays sharp and precise. Pen’s unique, long leg lays flat against rulers and flexible curves for accurate alignment. Pens are perfectly balanced to comfortably fit in your hand. Instant writing, long storage time, and dry-out prevention are ensured by an airtight ring seal in the cap. Pen responds the moment it is touched to paper.