This item is now discontinued. View the direct replacement: RK-36620-43 or browse similar items
Cole-Parmer SC-200-96 Sample Concentrator for 96-Well Plates
Representative image only.
Cole-Parmer Item # RK-36620-41
Quickly concentrate multiple samples in a block at once
  • Variable needle position suits various Dri-Block® inserts
  • Small size makes it ideal for use under a fume hood
  • Needles, block, and block heater sold separately
  • For 96-well plate
  • 3 year warranty
Accessories for your sample concentrator
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)20 7/8
  • Width (in)9 7/16
  • Depth (in)11 5/8
  • DescriptionSample Concentrator for 96-Well Plates
  • Warranty3 Years
3 Year Warranty
Includes
Gas reservoir and stand.
Key Features
  • Variable needle position suits various Dri-Block® inserts
  • Small size makes it ideal for use under a fume hood
  • Needles, block, and block heater sold separately
  • For 96-well plate
  • 3 year warranty
More About this Item
Concentrator blows an inert gas over the surface of your samples via stainless steel needles (not included). Blown gas in combination with the heat from the heater produce ideal conditions for fast efficient evaporation. Insert needles into a silicone membrane in any configuration. Fully adjustable gas reservoir provides accurate height control. Maximum vertical traveling distance is 32 cm. Maximum gas usage is 15 L/min. Order PTFE-coated needles for use with corrosive samples.
Please Note: Effective June 2022, the TechneSample Concentrator for 96-Well Plates Model number FSC496D has now been rebranded Cole-Parmer under Model number SC-200-96. All the same great features and specifications, just with a new and improved look.