This item is now discontinued. Please browse similar items
Sanitary Gasket, Platinum-Cured Silicone, Flanged, 1-1/2" Tri-Clamp®
Representative image only.
Mfr # 40RXPX-F-150 – Item # RK-30564-04
Hands-free installation for difficult-to-reach locations!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Material: Platinum-Cured Silicone
  • Clamp Size: 1.5"
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPlatinum-cured silicone
  • Connection TypeSanitary
  • Sanitary Gasket Size (in)1 1/2
  • DescriptionSanitary Gasket, Flanged, Platinum-cured Silicone, 1-1/2\" Clamp Size
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Material: Platinum-Cured Silicone
  • Clamp Size: 1.5"
  • 1 year warranty
More About this Item
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port—once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time.
Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).