This item is now discontinued. Please browse similar items
Cole-Parmer Essentials Sand for Stainless Steel Bath, 1 Lb
Representative image only.
Cole-Parmer StableTemp Item # RK-36600-63
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSand for Stainless Steel Bath, 1 Lb
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Stainless Steel Sand Baths are ideal for irregular vessels or to hold a variety of different size vessels at one time. Constructed of stainless steel for superior corrosion resistance. Designed to hold sand, stainless steel shot, or non-volatile liquids. Fits into the Cole Parmer Stable Temp Dry Block Heaters in place of a modular block. Select the appropriate capacity for the size block heater.
Sand for use in Cole Parmer StableTemp sand baths is an ideal solution for the sample that does not fit in to a modular block. Sand offers great temperature stability. Choose a 1 pound package for each size block capacity, i.e. a two block capacity sand bath will require 2 lbs of sand.