This item is now discontinued. Please browse similar items
Thermo Scientific RNase Surface Decontaminant; 250 mL Bottle
Representative image only.
Thermo Scientific Mfr # 7000 – Item # RK-25715-00
Fast, effective way to remove RNase and DNA contamination
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRNase surface decontaminant, 250 ml bottle
More About this Item
These surface decontaminants provide a safe, fast, and proven alternative to remove nuclease and DNA contamination from benchtops, instruments, pipettors, glass, and plasticware. Conventional methods to remove RNA and DNA contamination use a Diethylpyro-carbonate (DEPC) treatment, which is suspected to be carcinogenic and must be handled with gloves and under approved fume
hoods at all time.
RNase AWAY leaves your instruments and other surface areas residue-free. You can spray, wipe, or soak your labware with RNase AWAY and remain confident your work is free of nuclease and nucleic acid contamination. It comes in 250-mL bottle or convenient 475- mL spray bottle.