Cole-Parmer Vial Cap, Preassembled Screw Closure, Blue, PTFE/Silicone Liner, 9 mm; 100/Pk
Representative image only.
Cole-Parmer Item # RK-98722-12
Keep your samples safe with these vial accessories
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This includes the cap and septa preassembled together—vial sold separately
  • Limited Lifetime Warranty
PRODUCT OPTIONS
$58.20CAD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeScrew-Thread
  • Cap Diameter (mm)9
  • Cap colorBlue
  • DescriptionPreassembled Screw Closure and Septa, Blue, PTFE/Silicone Lined, 9 mm
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Select from a variety of caps, septa and inserts that fit your application and needs.
  • This includes the cap and septa preassembled together—vial sold separately
  • Limited Lifetime Warranty
More About this Item
Suitable for use with Cole-Parmer® Versi-Vials
Preassembled closure and septa come ready to use with Wide-Neck Vials 98722-12 and 98722-02. Designed to work in robotic arm autosamplers.