Cole-Parmer PolarSafe® Smart Cap for Storage Dewars; 30-35L
Representative image only.
Cole-Parmer Item # RK-44260-78
Enhance storage and monitoring your temperature-sensitive samples
  • Robust cryopreservation solutions for long-term storage and transport of biological samples
  • Optimize sample protection and transportation with these dewar accessories
  • Suitable for use with 30 to 35L aluminum alloy liquid nitrogen tanks with 125 mm opening port
PRODUCT OPTIONS
$2,593.17CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Neck Inside Diameter (ID) (in)5
  • Capacity30 to 35 L
  • DescriptionSmart Cap for Storage Dewars; 30-35L
Key Features
  • Robust cryopreservation solutions for long-term storage and transport of biological samples
  • Optimize sample protection and transportation with these dewar accessories
  • Suitable for use with 30 to 35L aluminum alloy liquid nitrogen tanks with 125 mm opening port
More About this Item
These Cole-Parmer® Polarsafe® dewar accessories are compatible with Cole-Parmer® PolarSafe® Cryogenic Storage and Transport Dewars. Choose from a wide range of accessories including Smart Caps, trolleys, dewar monitor, and level rulers.
Choose accessories that are compatible with the size of dewar you have. These accessories can be used with Cryogenic Storage and Transport Dewars, Cryogenic Storage Dewars with Racks, and Dewar and Traceable® Temperature Data Logger Bundles.
SmartCap is a low energy consumption IoT-based module that can monitor liquid nitrogen level and temperature in a liquid nitrogen tank. The built-in high efficiency lithium battery can operate the device for more than two years.
Available for sale in the United States and Canada only.