This item is now discontinued. Please browse similar items
Cole-Parmer Pipette Tip Box, PP, with green Rack for 300 µl Tips
Representative image only.
Cole-Parmer Item # RK-25712-94
Ideal for pipette tip protection and storage
  • Hinged lid provides fast, safe pipette tip access
  • Tip rack layout meets ANSI SLAS 1-2004
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")4 3/4
  • Width (InchShort)3 1/2
  • Height (in)2 19/32
  • Max Volume (µL)300
  • Body MaterialPP
  • DescriptionPipette Tip Box, PP, with green rack for 300 µl tips
Key Features
  • Hinged lid provides fast, safe pipette tip access
  • Tip rack layout meets ANSI SLAS 1-2004
More About this Item
Each pipette tip storage box contains a hinged lid for easy one-hand opening, and a grid insert for quick organization. Each tip box can safely house a variety of pipette tip brands.
The transparent polycarbonate boxes featuring a 128-place grid insert, are designed for autoclaving tips at 121°C. They are 100% form stable, even after numerous autoclave cycles. Two sizes are available; one to accommodate 200 μL pipette tips and the other for 1000 μL pipette tips.
The polypropylene boxes are available in four sizes to accommodate either 20-, 200-, 300-, or 1000-µl pipette tips. Each box comes with a 96-place color coded grid insert.