Cole-Parmer pH reference half-cell, Ag/AgCl fill, glass body
Representative image only.
Cole-Parmer Item # RK-05990-60
For research measurements
  • Glass material
  • Half-cell
  • 6 month warranty
$169.44CAD / Each
BUY MORE & SAVE
  • 1-3$169.44CAD / Each
  • 4+$152.50CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsResearch measurements
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorReference pin
  • Diameter (mm)12
  • Length (mm)154
  • DescriptionpH reference half-cell, Ag/AgCl fill, glass body
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Glass material
  • Half-cell
  • 6 month warranty
More About this Item
Use to build a pH electrochemical cell.