This item is now discontinued. Please browse similar items
Replacement General Purpose Penetration Probe, Stainless Steel Stem; 6" Length
Representative image only.
Traceable Item # RK-90080-13
Optimize performance of your thermometer
  • Probe is submersible up to the handle for ideal measuring
  • Features a temperature range of –58 to 302°F (–50 to 150°C)
  • Probe has 0.1 cm diameter, 7 cm stem length, 14 cm overall length, and 152 cm cable length
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPenetration Probe for Scientific Thermistor Thermometer 90080-12
  • Warranty1 Year
1 Year Warranty
Key Features
  • Probe is submersible up to the handle for ideal measuring
  • Features a temperature range of –58 to 302°F (–50 to 150°C)
  • Probe has 0.1 cm diameter, 7 cm stem length, 14 cm overall length, and 152 cm cable length
  • 1 year warranty
More About this Item
Compatible for use with the Traceable Scientific Thermistor Thermometer 90080-12, meter sold separately.