Cole-Parmer DLF-200 PCR Verticular Laminar Flow Cabinet, 48"; 115 VAC, 60 Hz

Representative image only.
Cole-Parmer Item # RK-78900-44
Protect your PCR samples against cross-contamination
  • Built-in germicidal UV lamp permits decontamination
  • Verticular laminar airflow with HEPA filtration
  • Innovative filtration technology
  • Produces ISO Class 5 compliant clean air
  • Meets OSHA, ANSI, and other international standards
  • Requires two pre-filters and two main HEPA filters
  • 3 year warranty

PRODUCT OPTIONS

$5,841.99CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Blower Typeebm-papst centrifugal fan
  • LightsLED & 253.7 nm 15 W UV lamp
  • Exterior Width (in)48
  • Exterior Height (in)35
  • Exterior Depth (in)27
  • Interior Width (in)48
  • Interior Height (in)24
  • Interior Depth (in)26.76
  • Power (VAC)115
  • Power (Hz)60
  • Shipping Weight (lbs)225
  • Shipping Weight (kg)102
  • Filter TypeHEPA main filter, polyester fiber pre-filter
  • DescriptionPCR Verticular Laminar Blow Cabinet, 48\"; 115 VAC, 60 Hz
  • Warranty3 Years
3 Year WarrantyUL Listed.

Includes

Power cord, polypropylene spill tray (78900-11), two main HEPA filters (78900-15), two pre-filters

Key Features

  • Built-in germicidal UV lamp permits decontamination
  • Verticular laminar airflow with HEPA filtration
  • Innovative filtration technology
  • Produces ISO Class 5 compliant clean air
  • Meets OSHA, ANSI, and other international standards
  • Requires two pre-filters and two main HEPA filters
  • 3 year warranty

More About this Item

The Cole-Parmer ductless PCR Worksation is the perfect cabinet for nonhazardous applications where biological or biohazard by-products are not generated, and user protection is not required. The cabinet provides a contaminant-free interior with a built-in germicidal 253.7 nm UV lamp. The lamp has a timer to permit decontamination between cycles—set timer up to 60 minutes or to hold indefinitely.

Innovative filtration technology consists of a pre-filter and main HEPA filter which is 99.99% effective for 0.3 micron particulates. The filter chamber prevents contaminated air from encountering internal cabinet components, prolonging the lifespan. A filter door lock prevents unauthorized removal or accidental exposure to dirty filters.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat