This item is now discontinued. We recommend the following as suggested replacements: RK-24501-03
Cole-Parmer Omega® Fixed-Volume Pipettor, 10 µL; 1/EA
Representative image only.
Cole-Parmer Item # RK-24501-12
Ergonomic finger grip for comfortable use
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Volume: 10 µL
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes
  • Max Volume (µL)10
  • Accuracy (%)0.8
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeYellow
  • DescriptionFixed-Volume Pipettor, 10 µL; 1/EA
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO 9001:2008 Certified Supplier
Key Features
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Volume: 10 µL
  • 3 year warranty
More About this Item
These pipettors are offered in a wide-range of fixed volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.