This item is now discontinued. Please browse similar items
Cole-Parmer NP 74-2302 Single plate rotator for 96-well dialyzers
Representative image only.
Cole-Parmer Item # RK-02896-64
Keep you sample in constant motion
  • 139 µL swept volume
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSingle plate rotator for 96-well dialyzers
  • Warranty1 year
1 Year Warranty
Key Features
  • 139 µL swept volume
  • 1 year warranty
More About this Item
Rotator required for 96-well dialyzers. Features variable rotation rates; keeps sample in constant motion to speed up equilibrium dialysis. Ensures higher reproducibility.
Compatible with dialyzers 02896-70 and -72.