This item is now discontinued. Please browse similar items
Cole-Parmer Microscope Slides; frosted; one gross (144 slides)
Representative image only.
Cole-Parmer Item # RK-48501-00
Won’t stick or scratch
  • Easy to write on
  • Manufactured in accordance with ISO 8037/1
  • Measure 26 x 76 mm and 1 mm thick
  • Tolerance for thickness: ±0.05 mm
  • Planarity under 30 µm measured over 60 mm length
  • Choose from plain, frosted, or pink color frost options
SPECS & DESCRIPTION
Specifications & Description
  • Slide TypeFrosted
  • DescriptionMicroscope Slides; frosted; one gross (144 slides)
Key Features
  • Easy to write on
  • Manufactured in accordance with ISO 8037/1
  • Measure 26 x 76 mm and 1 mm thick
  • Tolerance for thickness: ±0.05 mm
  • Planarity under 30 µm measured over 60 mm length
  • Choose from plain, frosted, or pink color frost options
More About this Item
Our slides feature precision ground edges on all four sides. Use plain microscope slides in general purpose applications. The raised coating on the Colorfrost slides keeps them separated to prevent sticking and scratching and provides a smooth writing surface for easy labeling. Color frosted slides make identification a snap. Manufactured from extra-white soda-line glass with very low iron content.
Our slides feature precision ground edges on all four sides. Frosted slides allow easy marking of the slide for specimen identification. Slides measure 75 x 25 mm and are 1 mm thick.