This item is now discontinued. Please browse similar items

Cole-Parmer Horizontal Mini-gel unit, 7.5 x 10 cm gel bed

Representative image only.
Cole-Parmer Item # RK-28559-10
Compact, leak-free casting—ideal for quick screening
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)250
  • Gel Width (cm)7.5
  • Gel Length (cm)10
  • Cell Width (in)3.88
  • Cell Height (in)4
  • Cell Depth (in)7.88
  • Cell Width (cm)9.86
  • Cell Height (cm)10.16
  • Cell Depth (cm)20.02
  • DescriptionHorizontal Mini-gel unit, 7.5 x 10 cm gel bed
  • Warranty3 Years
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

Gel beds and trays are UV-transparent acrylic—scan gels at 275 to 325 nm without removing them from the unit. The tapered baffle design eliminates the need for sealing or taping. Compact format saves time and costly reagents. Optional gel trays and casters let you pour six gels simultaneously. Order combs, gel trays, and gel casters separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat