This item is now discontinued. Please browse similar items
Cole-Parmer High Speed Bottle Top Filtration Pedestal Stand
Representative image only.
Cole-Parmer Item # RK-06060-95
Compatible with Cole-Parmer high speed bottle top filtrations
SPECS & DESCRIPTION
Specifications & Description
  • ConnectionsVacuum hose
  • ColorBlack
  • DescriptionBottle Top Filtration, Pedestal Stand
More About this Item
The optional pedestal or cradle ring to create a convenient docking station for hands-free filtering.
The pedestal stand provides a permanent vacuum connection along with a font on/off control knob.
The cradle ring attaches to any standard lab stand and features a side on/off control knob.