Cole-Parmer Grade 55 Hardened-Ashless Filter Paper, 9 cm Diameter; 100/PK
Representative image only.
Cole-Parmer Item # RK-06648-68
Fast-filtering grade is ideal for gravimetric analysis of coarse particles
  • Suited to collecting wet precipitates
  • Tough, smooth surface free of loose fibers
  • Great for filtering chloride and phosphorus in cement, coal and coke
  • 9 cm diameter
  • 6 month warranty
PRODUCT OPTIONS
$63.06CAD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCotton
  • Pore Size (micron)15
  • Diameter (cm)9
  • Retentive Pore Liquid (µm)15
  • DescriptionGrade 55 Hardened-Ashless Filter Paper, 9 cm Diameter; 100/Pk
  • Warranty6 month
6 Month WarrantyMade in the U.S.A.
Key Features
  • Suited to collecting wet precipitates
  • Tough, smooth surface free of loose fibers
  • Great for filtering chloride and phosphorus in cement, coal and coke
  • 9 cm diameter
  • 6 month warranty
More About this Item
Use these specially treated hardened-ashless filter papers for critical analytical techniques requiring increased wet-strength and handling capacity. These fast-filtering filter papers are ideal for gelatinous precipitates in acid and alkali solutions, as well as air and food analysis.