This item is now discontinued. Please browse similar items
Cole-Parmer Glass Plate Set for Dual Vertical Mini-Gel System Model 28575-00
Representative image only.
Cole-Parmer Item # RK-28575-90
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Glass Width (in)4.45
  • Glass Width (cm)11.3
  • Glass Height (cm)10
  • DescriptionGlass Plate Set for Dual Vertical Mini-Gel System Model 28575-00
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Glass Plate Sets consist of a notched plate and a back plate of 1/8'' soda lime glass. Radius-bottom corners on the back plate accept Gel Wrap™ gaskets for a leak-proof seal. All edges are sanded and polished.