This item is now discontinued. Please browse similar items
Cole-Parmer General Chemistry Laboratory Safety Kit
Representative image only.
Cole-Parmer Item # RK-25000-47
Great for high school and entry-level college chemistry courses!
SPECS & DESCRIPTION
Specifications & Description
  • SizeOne Size
  • DescriptionGeneral Chemistry Laboratory Safety Kit
Includes
More About this Item
Handy kit includes a lab notebook and scientific calculator to record and analyze experiment results, and safety apron and clear safety goggles to protect your clothes and eyes from chemical splashes.
The 184-page, permanently-bound, laboratory notebook features numbered pages printed with a green grid for plotting tables and graphs. Additional pages include an issuance page, instructions for keeping research records, and a table of contents. Record experiments in accordance with Good Documentation Practices (GDP) — numbered pages have spaces for signatures and dates. Uncoated Polypaper® is highly resistant to moisture, chemicals, tearing, and aging; works best with ballpoint pen or felt-tip marker.
The TI-36X Pro scientific calculator is solar powered and features a four-line display to show multiple calculations simultaneously or in tabular form. Choose from three solvers: numeric equation, polynomial, and system of linear equations. Select degrees/radians or floating/fix number format modes. Determine the numeric derivative and integral for real functions. Calculator includes a slide case and instructions.
The one-size-fits-most Ansell neoprene apron is 35 inches (89 cm) wide and 45 inches (114 cm) long. Neoprene aprons are rugged enough to withstand frequent use and resistant to a wide range of acids and oils. Extra-long ties and reinforced metal grommets prevent tearing. Wider bib style offers added chest protection.
Safety goggles feature indirect ventilation and an impact-resistant, low profile design for chemical splash protection. UVExtreme® coating for fog, scratch, and static resistance helps maintain clear vision, and the curved lens provides superior optics, peripheral vision, and comfort during long periods of use. Complies to ANSI Z87.1-2010, CSA Z94.3 standards.