This item is now discontinued. Please browse similar items
Cole-Parmer Gel cassette for Electrophoretic Blotting System 28580-20
Representative image only.
Cole-Parmer Item # RK-28580-54
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (cm)1
  • Width (cm)9
  • DescriptionGel cassette for Electrophoretic Blotting System 28580-20
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Gel cassettes consist of two hinged, perforated fiberglass panels and two Scotch Brite® pads. Place gel and membrane (nitrocellulose, nylon, or other) between pads to run transfer.