This item is now discontinued. Please browse similar items
Cole-Parmer Fold-Up Benchtop Magnifier, Illuminated, 0.5", 10x
Representative image only.
Cole-Parmer Item # RK-98766-68
Fold up to protect lens—and take them anywhere!
  • All-glass optic assures perfect focus with zero distortion
  • Choose from illuminated/nonilluminated models and the magnification power that suits inspection needs
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x
  • Viewing Area1/2\" diameter
  • LensSingle
  • Mounting TypeBench top/Handsfree
  • Light SourceLED
  • BatteryAA Battery
  • ColorBlack
  • DescriptionFold-Up Benchtop Magnifier, Illuminated, 0.5\", 10x
1 Year Warranty
Includes
Batteries.
Key Features
  • All-glass optic assures perfect focus with zero distortion
  • Choose from illuminated/nonilluminated models and the magnification power that suits inspection needs
More About this Item
These fold-up benchtop magnifiers are designed for use in quality control labs to examine food and electronics. Stand-alone units allow you to inspect with both hands. White ruler markings in both inches and millimeters on the lower panel assist in making measurements