This item is now discontinued. Please browse similar items
Cole-Parmer Filter Membrane, 47 mm, .45 µm, PES; 50/Pk
Representative image only.
Cole-Parmer Item # RK-06060-09
Economical, efficient surface filtering for the retention of particles or microorganisms
  • Materials adhere to USP Class VI Standards and FDA compliances
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPES
  • Pore Size (µm)0.45
  • Diameter (mm)47
  • DescriptionFilter Membrane, 47 mm, .45 µm, PES; 50/Pk
Sterile
Key Features
  • Materials adhere to USP Class VI Standards and FDA compliances
More About this Item
Nylon membranes offer good chemical resistance, making them ideal for filtering your more aggressive solutions such as alcohols and DMSO solutions.
Hydrophobic PTFE membranes are bonded to a polypropylene support for improved handling. They are highly compatible with almost all solvents including strong acids and bases.
Mixed cellulose ester (MCE) membranes are gridded for easy cell counts. These sterile disc filters are used for the microbiological analysis of water and other fluids in the food & beverage and pharmaceutical industries. Sterilized by gamma irradiation and individually packaged.