This item is now discontinued. Please browse similar items
Cole-Parmer Filter Breakthrough Indicator; 5 Pk
Representative image only.
Cole-Parmer Item # RK-06059-04
Know when to change your solvent waste system filter
  • Fits onto solvent waste system filters
  • Choose from six different pack sizes
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCole-Parmer Carboy Solvent Waste Systems
  • DescriptionFilter Breakthrough Indicator; 5/Pk
Key Features
  • Fits onto solvent waste system filters
  • Choose from six different pack sizes
More About this Item
Stay safe—indicator changes color to red when trap filters need to be replaced. Designed to indicate the presence of common HPLC solvent vapors like acetone, acetonitrile, chloroform, dichloromethane, ethanol, ethyl acetate, hexafluoroisopropanol, methanol, phenol, sulfolane, and tetrahydrofuran. Operating temperature: 4 to 35°C (39 to 95°C).