This item is now discontinued. Please browse similar items
Cole-Parmer Fermentation DO Probe; SS/Polarographic/420 mm, 3/4" NPT
Representative image only.
Cole-Parmer Item # RK-05726-04
Designed for fermentation applications where probes are repeatedly steam-sterilized
  • 316 SS Construction
  • Polarographic
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes and Sensors
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Min Temperature (° F)32
  • Max Temperature (° F)176
  • Temperature compensationAutomatic
  • Length (cm)42
  • DescriptionFermentation DO Probe; SS/Polarographic/420 mm, 3/4\" NPT
  • Warranty1 year
1 Year Warranty
Key Features
  • 316 SS Construction
  • Polarographic
  • 1 year warranty
More About this Item
All models feature a polarographic sensor, and are available in a variety of body diameters, mounting styles, and immersion lengths. Temperature compensated probes feature 100 Ω RTD temperature measuring sensors. Order cables separately.