This item is now discontinued. Please browse similar items
Cole-Parmer Fermentation DO Probe; Glass/Polarographic/300 mm
Representative image only.
Cole-Parmer Item # RK-05643-04
Low-cost, glass-body design
  • Polarographic
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes and Sensors
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Min Temperature (° F)32
  • Max Temperature (° F)140
  • Length (cm)30
  • DescriptionFermentation DO Probe; Glass/Polarographic/300 mm
  • Warranty1 year
1 Year Warranty
Key Features
  • Polarographic
  • 1 year warranty
More About this Item
These economical glass-body probes are ideal for your laboratory fermentation applications where sterilization takes place in a separate autoclave. Probes feature a threaded detachable cable lead. Order appropriate cable separately.