This item is now discontinued. Please browse similar items
Cole-Parmer Fermentation 3 or 5 Liter System Antifoam Probe; 10mm Thread/130mm Immersion depth
Representative image only.
Cole-Parmer Item # RK-01919-62
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFermentation 3 or 5 Liter System Antifoam Probe; 10mm Thread/130mm Immersion depth
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Order a probe according to your vessel size; all probes require cable 01919-65.