This item is now discontinued. Please browse similar items
Cole-Parmer Essentials Wall Clamp, Nickel-Plated Zinc, 0.2" to 0.39" Grip Range
Representative image only.
Cole-Parmer Essentials Item # RK-08027-00
Provide support for your specific laboratory equipment
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeFrame and Rod
  • Min Grip Range (in)0.2
  • Max Grip Range (in)0.39
  • Length (in)3.15
  • DescriptionWall Clamp, Nickel-Plated Zinc, 0.2\" to 0.39\" Grip Range
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Clamp has an integral self-tapping screw designed to mount directly into a wall on one end and a clamp on the other end for attaching to a frame rod.