Cole-Parmer Essentials Inoculating Streaker Loops, Small, Blue; 2.5 mm ID; 5/Pk
Representative image only.
Cole-Parmer Essentials Item # RK-04396-42
Nichrome wire composition cools quickly and is less expensive than platinum
  • Select, inoculate, or transfer microbiological samples
PRODUCT OPTIONS
$103.14CAD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Loop Inside Diameter (mm)2.5
  • ColorBlue
  • DescriptionInoculating Streaker Loops, Small, Blue; 2.5 mm ID; 5/Pk
Key Features
  • Select, inoculate, or transfer microbiological samples
More About this Item
Loops are made of high quality twisted nichrome wire and attached to a light-weight aluminum handle. Handles are insulated to protect the hand during use. Loops have been designed to slide along agar surfaces without cutting or tearing. Loops and Handles cool rapidly after sterilization and are offered calibrated (1µl & 10µl) or non-calibrated. Non-calibrated Loops and Handles are available as small, medium or large and are color coded for easy identification.