This item is now discontinued. We recommend the following as suggested replacements: RK-09968-00
Cole-Parmer Essentials Fine-Point Permanent Lab Markers, Red; 10/Box
Representative image only.
Cole-Parmer Essentials Item # RK-09964-20
Quick-drying ink permanently marks plastic
  • Washes off glass, metal, or porcelain
  • Won’t bleed when subjected to water or alcohol
SPECS & DESCRIPTION
Specifications & Description
  • ColorRed
  • DescriptionFine-Point Permanent Lab Markers, Red; 10/Box
Key Features
  • Washes off glass, metal, or porcelain
  • Won’t bleed when subjected to water or alcohol
More About this Item
Fine-point fiber tip writes on cold, wet surfaces.